Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766376.1 | XP_008764598.1 | 9 - 10 | 7781 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |