Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025010.1 | NP_001020181.1 | 8 - 9 | 2346 | 71 | |
GenBank mRNA | BC089834.1 | - | 8 - 9 | 2346 | 7325650 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |