Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014089.1 | NP_001014111.1 | 5 - 6 | 928 | 122 | |
XM_006242859.2 | XP_006242921.1 | 5 - 6 | 3575 | 122 | ||
XM_008766134.1 | XP_008764356.1 | 6 - 7 | 3538 | 122 | ||
GenBank mRNA | BC087163.1 | - | 5 - 6 | 928 | 7135124 | 122 |
FQ230557.1 | - | 3 - 4 | 272 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |