Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766431.1 | XP_008764653.1 | 7 - 8 | 1964 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |