Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104640.1 | NP_001098110.1 | 4 - 5 | 572 | 64 | |
XM_006256574.2 | XP_006256636.1 | 4 - 5 | 639 | 64 | ||
GenBank mRNA | DQ503410.1 | - | 4 - 5 | 572 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |