Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108841.1 | NP_001102311.1 | 729 | 115 | ||
XM_008768584.1 | XP_008766806.1 | 3 - 4 | 338 | 115 | ||
XM_008768585.1 | XP_008766807.1 | 3 - 4 | 333 | 115 | ||
XM_008768586.1 | XP_008766808.1 | 3 - 4 | 333 | 115 | ||
XM_008768587.1 | XP_008766809.1 | 3 - 4 | 338 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |