Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107117.1 | NP_001100587.1 | 9 - 10 | 923 | 61 | |
XM_006248804.2 | XP_006248866.1 | 8 - 9 | 1039 | 61 | ||
XM_006248805.2 | XP_006248867.1 | 8 - 9 | 941 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |