Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_224885.7 | XP_224885.3 | 6 - 7 | 1361 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |