Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134880.1 | NP_001128352.1 | 2 - 3 | 283 | 97 | |
GenBank mRNA | FQ209946.1 | - | 2 - 3 | 284 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |