Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134595.2 | NP_001128067.2 | 7 - 8 | 1232 | 88 | |
XM_006241491.2 | XP_006241553.1 | 7 - 8 | 2878 | 88 | ||
GenBank mRNA | FQ211772.1 | - | 7 - 8 | 1168 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |