Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014202.1 | NP_001014224.1 | 1 - 2 | 196 | 87 | |
GenBank mRNA | BC085877.1 | - | 1 - 2 | 196 | 7191715 | 87 |
FQ218639.1 | - | 1 - 2 | 233 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |