Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130582.2 | NP_001124054.2 | 6 - 7 | 1017 | 72 | |
XM_006242121.2 | XP_006242183.1 | 5 - 6 | 834 | 72 | ||
GenBank mRNA | BC168759.1 | - | 6 - 7 | 1017 | 6888763 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |