Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750338.3 | XP_003750386.1 | 9 - 10 | 874 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |