Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014085.1 | NP_001014107.1 | 1 - 2 | 305 | 66 | |
XM_006241811.1 | XP_006241873.1 | 1 - 2 | 349 | 66 | ||
XM_006241812.1 | XP_006241874.1 | 1 - 2 | 349 | 66 | ||
GenBank mRNA | BC083712.1 | - | 1 - 2 | 305 | 7190634 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |