Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012174.1 | NP_001012174.1 | 3 - 4 | 365 | 94 | |
XM_006256222.2 | XP_006256284.1 | 3 - 4 | 1114 | 94 | ||
XM_006256224.2 | XP_006256286.1 | 4 - 5 | 1181 | 94 | ||
GenBank mRNA | BC085868.1 | - | 3 - 4 | 365 | 7191081 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |