Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107618.1 | NP_001101088.1 | 11 - 12 | 2586 | 88 | |
XM_006256207.2 | XP_006256269.1 | 11 - 12 | 3158 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |