Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-DOb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255970.2 | XP_006256032.1 | 5 - 6 | 908 | 88 | |
GenBank mRNA | KC222930.1 | - | 5 - 6 | 706 | 88 | |
KC222931.1 | - | 5 - 6 | 706 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |