Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107617.2 | NP_001101087.2 | 7 - 8 | 1063 | 91 | |
XM_006256205.2 | XP_006256267.1 | 7 - 8 | 1109 | 91 | ||
XM_008772729.1 | XP_008770951.1 | 7 - 8 | 1109 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |