Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ang2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251898.2 | XP_006251960.1 | 1 - 3 | 371 | 65 |
Gene Symbol: Ang | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251874.2 | XP_006251936.1 | 1 - 2 | 162 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |