Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191107.1 | NP_001178036.1 | 4 - 5 | 302 | 86 | |
XM_006239995.2 | XP_006240057.1 | 4 - 5 | 551 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |