Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145449.1 | NP_001138921.1 | 1 - 2 | 92 | 89 | |
GenBank mRNA | CB765303.1 | - | 1 - 2 | 92 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |