Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199237.1 | NP_954707.1 | 3 - 4 | 370 | 72 | |
GenBank mRNA | L02842.1 | - | 3 - 4 | 370 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |