Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729615.3 | XP_002729661.1 | 9 - 10 | 1290 | 88 | |
XM_006239869.2 | XP_006239931.1 | 9 - 10 | 1317 | 88 | ||
XM_006239870.2 | XP_006239932.1 | 8 - 9 | 1191 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |