Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007013.1 | NP_001007014.1 | 1 - 2 | 202 | 101 | |
GenBank mRNA | AY437504.1 | - | 1 - 2 | 202 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |