Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106740.1 | NP_001100210.1 | 1 - 2 | 524 | 70 | |
XM_006240222.2 | XP_006240284.1 | 1 - 2 | 574 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Rattus norvegicus Probe spans exons |