Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006980.1 | NP_001006981.1 | 2 - 3 | 220 | 135 | |
XM_006239982.2 | XP_006240044.1 | 3 - 4 | 341 | 135 | ||
GenBank mRNA | BC083576.1 | - | 2 - 3 | 220 | 7113186 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |