Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009542.2 | NP_001009542.1 | 2 - 3 | 297 | 85 | |
XM_006232501.2 | XP_006232563.1 | 2 - 3 | 258 | 85 | ||
GenBank mRNA | BC067245.1 | - | 2 - 3 | 301 | 6888212 | 85 |
FQ213019.1 | - | 2 - 3 | 298 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |