Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: She | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062249.5 | XP_001062249.1 | 1 - 2 | 787 | 76 | |
XM_008761244.1 | XP_008759466.1 | 1 - 2 | 787 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |