Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251471.2 | XP_006251533.1 | 8 - 9 | 1031 | 80 | |
XM_008770406.1 | XP_008768628.1 | 8 - 9 | 4613 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |