Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191728.1 | NP_001178657.1 | 2 - 3 | 694 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |