Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126270.1 | NP_001119742.1 | 2 - 3 | 322 | 91 | |
XM_006245657.2 | XP_006245719.1 | 11 - 12 | 1360 | 91 | ||
XM_008767423.1 | XP_008765645.1 | 11 - 12 | 1372 | 91 | ||
GenBank mRNA | BC161935.1 | - | 11 - 12 | 1307 | 9022458 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |