Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126270.1 | NP_001119742.1 | 5 - 6 | 659 | 64 | |
XM_006245657.2 | XP_006245719.1 | 14 - 15 | 1697 | 64 | ||
XM_008767423.1 | XP_008765645.1 | 14 - 15 | 1709 | 64 | ||
GenBank mRNA | BC161935.1 | - | 14 - 15 | 1644 | 9022458 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |