Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750698.3 | XP_003750746.1 | 9 - 10 | 5628 | 67 | |
XM_006244936.2 | XP_006244998.1 | 9 - 10 | 2564 | 67 | ||
XM_006244937.2 | XP_006244999.1 | 10 - 11 | 2552 | 67 | ||
XM_006244938.2 | XP_006245000.1 | 9 - 10 | 3582 | 67 | ||
XM_008767152.1 | XP_008765374.1 | 9 - 10 | 3140 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Rattus norvegicus 1700 Probe spans exons |