Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blmh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034163.1 | NP_001029335.1 | 10 - 11 | 1322 | 80 | |
GenBank mRNA | BC128701.1 | - | 10 - 11 | 1207 | 7379958 | 80 |
CB583895.1 | - | 4 - 5 | 458 | 80 | ||
D87336.1 | - | 10 - 11 | 1225 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |