Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114606.1 | NP_001108078.1 | 8 - 9 | 1488 | 127 | |
XM_006246509.2 | XP_006246571.1 | 8 - 9 | 3085 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |