Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106253.1 | NP_001099723.1 | 3 - 4 | 499 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |