Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106253.1 | NP_001099723.1 | 4 - 5 | 646 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |