Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024901.1 | NP_001020072.1 | 2 - 3 | 553 | 88 | |
XM_008773422.1 | XP_008771644.1 | 2 - 3 | 655 | 88 | ||
GenBank mRNA | DQ058659.1 | - | 2 - 3 | 553 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |