Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_236641.6 | XP_236641.6 | 4 - 5 | 417 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |