Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233425.2 | XP_006233487.1 | 8 - 9 | 1518 | 91 | |
XM_008761538.1 | XP_008759760.1 | 9 - 10 | 1713 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |