Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229523.2 | XP_006229585.1 | 4 - 5 | 798 | 87 | |
XM_006229524.2 | XP_006229586.1 | 4 - 5 | 813 | 87 | ||
XM_006229527.2 | XP_006229589.1 | 4 - 5 | 418 | 87 | ||
XM_006229528.2 | XP_006229590.1 | 4 - 5 | 780 | 87 | ||
XM_006229529.2 | XP_006229591.1 | 4 - 5 | 795 | 87 | ||
XM_219738.8 | XP_219738.6 | 3 - 4 | 957 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |