Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025118.1 | NP_001020289.1 | 8 - 9 | 1513 | 77 | |
XM_006232886.2 | XP_006232948.1 | 11 - 12 | 2737 | 77 | ||
XM_008761304.1 | XP_008759526.1 | 12 - 13 | 2695 | 77 | ||
GenBank mRNA | BC091386.1 | - | 8 - 9 | 1513 | 7321358 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |