Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106339.1 | NP_001099809.1 | 2 - 3 | 262 | 73 | |
XM_006231087.2 | XP_006231149.1 | 3 - 4 | 431 | 73 | ||
XM_006231088.2 | XP_006231150.1 | 3 - 4 | 431 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |