Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012696.2 | NP_036828.2 | 997 | 127 | ||
GenBank mRNA | BC087028.1 | - | 997 | 7130388 | 127 | |
FQ211035.1 | - | 405 | 129 | |||
FQ219050.1 | - | 403 | 127 | |||
M11883.1 | - | 767 | 127 | |||
X02299.1 | - | 732 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |