Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014024.1 | NP_001014046.1 | 1 - 2 | 385 | 127 | |
GenBank mRNA | BC079355.1 | - | 1 - 2 | 385 | 7108257 | 127 |
EU644751.1 | - | 1 - 2 | 232 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |