Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759213.1 | XP_008757435.1 | 4 - 5 | 1824 | 160 | |
XM_008759214.1 | XP_008757436.1 | 4 - 5 | 1341 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |