Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033957.1 | NP_001029129.1 | 1 - 2 | 665 | 63 | |
GenBank mRNA | AF454549.1 | - | 1 - 2 | 849 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Rattus norvegicus Probe spans exons |