Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000937.1 | NP_001000937.1 | 64 | 103 |
Gene Symbol: LOC100909935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748915.2 | XP_003748963.1 | 1 | 78 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |