Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203470.1 | NP_982296.1 | 1 | 563 | 76 | |
XM_008760116.1 | XP_008758338.1 | 2 | 787 | 76 | ||
XM_008760117.1 | XP_008758339.1 | 2 | 669 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |