Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765394.1 | XP_008763616.1 | 1 | 684 | 178 | |
XM_008765395.1 | XP_008763617.1 | 1 | 684 | 178 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |